3,5-Dichlorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

008033-1G 008033-5G
FLUO008033-1GEA 0 GBP
FLUO008033-1G FLUO008033-5G
3,5-Dichlorobenzaldehyde
3,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 215…222 °C (7 mmHg)
Melting Pt: 63
Storage Temperature: Ambient
MDL Number: MFCD00003352
CAS Number: 10203-08-4
EINECS: 233-499-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR