3,4-Dihydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Protocatechuic acid
37580-100G-F
37580-25G-F
SIAL37580-100G-FEA
89.6
GBP
SIAL37580-100G-F
SIAL37580-25G-F
3,4-Dihydroxybenzoic acid, Sigma-Aldrich®
3,4-Dihydroxybenzoic acid
Formula:
C₇H₆O₄ Boiling Pt: 125…135 °C (0.001 mmHg) Melting Pt: 202…204 °C Density: 1.5415 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002509 CAS Number: 99-50-3 EINECS: 202-760-0 Merck Index: 12,08080 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...