5-Nitroanthranilic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Amino-5-nitrobenzoic acid

018896-10G 018896-100G 018896-25G
FLUO018896-10GEA 0 GBP
FLUO018896-10G FLUO018896-100G FLUO018896-25G
5-Nitroanthranilic acid
5-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
Melting Pt: 278 °C
Storage Temperature: Ambient
MDL Number: MFCD00017039
CAS Number: 616-79-5
EINECS: 210-493-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR