4-Nitroanthranilic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Amino-4-nitrobenzoic acid

011363-100G 011363-25G
FLUO011363-100GEA 0 GBP
FLUO011363-100G FLUO011363-25G
4-Nitroanthranilic acid
4-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
Melting Pt: 271 °C
MDL Number: MFCD00007262
CAS Number: 619-17-0
EINECS: 210-583-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR