3-Ethoxybenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018662-2G 018662-10G
FLUO018662-2GEA 0 GBP
FLUO018662-2G FLUO018662-10G
3-Ethoxybenzonitrile
3-Ethoxybenzonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00001805
CAS Number: 25117-75-3
EINECS: 246-632-2
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR