4-Aminobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
024179-100G
024179-25G
FLUO024179-100GEA
0
GBP
FLUO024179-100G
FLUO024179-25G
4-Aminobenzonitrile
4-Aminobenzonitrile
Formula:
C₇H₆N₂ Boiling Pt: 167 °C (1.1 mmHg) Melting Pt: 84…87 °C Density: 1.274 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007821 CAS Number: 873-74-5 EINECS: 212-850-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...