4-Hydroxy-3-nitrobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018721-10G 018721-25G
FLUO018721-10GEA 0 GBP
FLUO018721-10G FLUO018721-25G
4-Hydroxy-3-nitrobenzaldehyde
4-Hydroxy-3-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00007117
CAS Number: 3011-34-5
EINECS: 221-141-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR