2-Bromothiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-Bromothiophenol, 2-Bromobenzene-1-thiol, 2-Bromobenzenethiol, 1-Bromo-2-mercaptobenzene

001577-5G 001577-25G 001577-100G
FLUO001577-5GEA 0 GBP
FLUO001577-5G FLUO001577-25G FLUO001577-100G
2-Bromothiophenol
2-Bromothiophenol
Formula: C₆H₅BrS
Boiling Pt: 128 °C (1013 hPa)
Density: 1.604 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00004827
CAS Number: 6320-02-1
EINECS: 228-665-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR