4-Bromoveratrole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001141-100G
001141-25G
FLUO001141-100GEA
0
GBP
FLUO001141-100G
FLUO001141-25G
4-Bromoveratrole
4-Bromoveratrole
Formula:
C₈H₉BrO₂ Boiling Pt: 255 °C (1013 hPa) Density: 1.5068 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008381 CAS Number: 2859-78-1 EINECS: 220-677-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...