3-Bromoanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003058-100G 003058-250G 003058-1KG
FLUO003058-100GEA 0 GBP
FLUO003058-100G FLUO003058-250G FLUO003058-1KG
3-Bromoanisole
3-Bromoanisole
Formula: C₇H₇BrO
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1,49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR