1-Bromo-2-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002709-100G
002709-25G
FLUO002709-100GEA
0
GBP
FLUO002709-100G
FLUO002709-25G
1-Bromo-2-nitrobenzene
1-Bromo-2-nitrobenzene
Formula:
C₆H₄BrNO₂ Boiling Pt: 261 °C (1013 hPa) Melting Pt: 39…42 °C Density: 1.62 g/cm³ (20 °C) Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007045 CAS Number: 577-19-5 EINECS: 209-409-0 UN: 3459 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...