4-Bromoaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
002472-250G
002472-1KG
FLUO002472-250GEA
0
GBP
FLUO002472-250G
FLUO002472-1KG
4-Bromoaniline
4-Bromoaniline
Formula:
C₆H₆BrN MW: 172.02 g/mol Boiling Pt: 223 Melting Pt: 62…64 °C Density: 1.799 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007822 CAS Number: 106-40-1 EINECS: 203-393-9 UN: 2811 ADR: 6.1 III Merck Index: 13,01388 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...