1-Bromo-3-iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromoiodobenzene
001125-100G
001125-25G
FLUO001125-100GEA
0
GBP
FLUO001125-100G
FLUO001125-25G
1-Bromo-3-iodobenzene
1-Bromo-3-iodobenzene
Formula:
C₆H₄BrI MW: 282.91 g/mol Boiling Pt: 240 °C (1013 hPa) Melting Pt: –9.3 °C Density: 2,21 g/cm³ (20 °C) |
MDL Number:
MFCD00001043 CAS Number: 591-18-4 EINECS: 209-703-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...