2-Bromo-5-fluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-4-fluoro-2-hydroxybenzene

004232-25G 004232-100G
FLUO004232-25GEA 0 GBP
FLUO004232-25G FLUO004232-100G
2-Bromo-5-fluorophenol
2-Bromo-5-fluorophenol
Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 184…186 °C (1013 hPa)
Density: 1,717 g/cm³ (20 °C)
MDL Number: MFCD00040939
CAS Number: 147460-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR