4-Bromo-2-fluorophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
004233-100G
004233-25G
FLUO004233-100GEA
0
GBP
FLUO004233-100G
FLUO004233-25G
4-Bromo-2-fluorophenol
4-Bromo-2-fluorophenol
Formula:
C₆H₄BrFO Boiling Pt: 90 °C (15 mmHg) Density: 1,74 g/cm³ (20 °C) |
MDL Number:
MFCD00011722 CAS Number: 2105-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...