1-Bromo-4-(trifluoromethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001402-25G 001402-100G 001402-250G
FLUO001402-25GEA 0 GBP
FLUO001402-25G FLUO001402-100G FLUO001402-250G
1-Bromo-4-(trifluoromethoxy)benzene
Formula: C₇H₄BrF₃O
Boiling Pt: 153 °C (1013 hPa)
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040834
CAS Number: 407-14-7
EINECS: 206-979-2
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR