1-Bromo-3-(trifluoromethoxy)benzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001480-25G
001480-100G
FLUO001480-25GEA
0
GBP
FLUO001480-25G
FLUO001480-100G
1-Bromo-3-(trifluoromethoxy)benzene
1-Bromo-3-(trifluoromethoxy)benzene
Formula:
C₇H₄BrF₃O Boiling Pt: 78…81 °C (55 mmHg) Density: 1.635 g/cm³ (20 °C) |
MDL Number:
MFCD00040943 CAS Number: 2252-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...