1-Bromo-4-fluoro-2-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010673-1G 010673-5G 010673-25G
FLUO010673-1GEA 0 GBP
FLUO010673-1G FLUO010673-5G FLUO010673-25G
1-Bromo-4-fluoro-2-nitrobenzene
1-Bromo-4-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MDL Number: MFCD00055530
CAS Number: 446-09-3
EINECS: 207-160-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR