Cholecalciferol (Vitamin D3), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Vitamin D3, Colecalciferol
ampouled under argon
C9774-5AMP
C9774-1AMP
SIALC9774-5AMPEA
689
GBP
SIALC9774-5AMP
SIALC9774-1AMP
Cholecalciferol (Vitamin D3), Sigma-Aldrich®
Cholecalciferol (Vitamin D3)
Formula:
C₂₇H₄₄O |
MDL Number:
MFCD00078131 CAS Number: 67-97-0 EINECS: 200-673-2 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...