D-Cycloserine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(R)-(+)-Cycloserine
, D-4-Aminoisoxazolidin-3-one
, Cycloserine
, D(+)-Cycloserine
, (R)-4-Aminoisoxazolidin-3-one
M-Clarity™ quality level = MQ200
SIALC6880-5GEA
476
GBP
SIALC6880-5G
SIALC6880-1G
D-Cycloserine, Sigma-Aldrich®
D-Cycloserine
Formula:
C₃H₆N₂O₂ MW: 102.09 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00005353 CAS Number: 68-41-7 EINECS: 200-688-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...