D-Cycloserine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine

M-Clarity™ quality level = MQ200

C3909-100MG C3909-250MG
SIALC3909-100MGEA 433 GBP
SIALC3909-100MG SIALC3909-250MG
D-Cycloserine, Sigma-Aldrich®
D-Cycloserine
Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR