Triisopropanolamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine
M-Clarity™ quality level = MQ100
254746-1KG
254746-100G
SIAL254746-1KGEA
63
GBP
SIAL254746-1KG
SIAL254746-100G
Triisopropanolamine, Sigma-Aldrich®
Triisopropanolamine
Formula:
C₉H₂₁NO₃ |
MDL Number:
MFCD00004533 CAS Number: 122-20-3 EINECS: 204-528-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...