3-Bromophenyl methyl sulfone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-3-(methylsulphonyl)benzene

019283-1G 019283-25G 019283-5G
FLUO019283-1GEA 0 GBP
FLUO019283-1G FLUO019283-25G FLUO019283-5G
3-Bromophenyl methyl sulfone
3-Bromophenyl methyl sulfone
Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
MDL Number: MFCD03095326
CAS Number: 34896-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR