2-Fluorophenyl methyl sulfone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Fluoro-2-(methylsulphonyl)benzene

019274-25G 019274-5G
FLUO019274-25GEA 0 GBP
FLUO019274-25G FLUO019274-5G
2-Fluorophenyl methyl sulfone
2-Fluorophenyl methyl sulfone
Formula: C₇H₇FO₂S
MW: 174.2 g/mol
MDL Number: MFCD03094199
CAS Number: 654-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR