3,5-Dichlorophenylhydrazinium chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018533-10G 018533-5G
FLUO018533-10GEA 0 GBP
FLUO018533-10G FLUO018533-5G
3,5-Dichlorophenylhydrazinium chloride
3,5-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MDL Number: MFCD00012938
CAS Number: 63352-99-8
EINECS: 264-109-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR