4-Chlorophenylhydrazinium chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008962-10G 008962-50G
FLUO008962-10GEA 0 GBP
FLUO008962-10G FLUO008962-50G
4-Chlorophenylhydrazinium chloride
4-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR