3-Fluorophenylhydrazinium chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Fluorphenyl)hydrazin HCl, (3-Fluorophenyl)hydrazine hydrochloride, 3-Fluorophenylhydrazine hydrochloride, 1-(3-Fluorophenyl)hydrazine hydrochloride

006333-5G 006333-25G 006333-100G
FLUO006333-5GEA 0 GBP
FLUO006333-5G FLUO006333-25G FLUO006333-100G
3-Fluorophenylhydrazinium chloride
3-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
MDL Number: MFCD00012934
CAS Number: 2924-16-5
EINECS: 220-886-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR