3,5-Bis(trifluoromethyl)benzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006015-1G 006015-5G
FLUO006015-1GEA 0 GBP
FLUO006015-1G FLUO006015-5G
3,5-Bis(trifluoromethyl)benzophenone
3,5-Bis(trifluoromethyl)benzophenone
Formula: C₁₅H₈F₆O
MDL Number: MFCD00015510
CAS Number: 21221-93-2
EINECS: 244-282-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR