5-Fluoro-2-methylbenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Fluoro-2-methylphenyl)methanol

005795-1G 005795-5G 005795-10G
FLUO005795-1GEA 0 GBP
FLUO005795-1G FLUO005795-5G FLUO005795-10G
5-Fluoro-2-methylbenzyl alcohol
5-Fluoro-2-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD01631527
CAS Number: 22062-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR