Cyclopropyl methyl ketone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-Cyclopropylethanone
005112-250G
005112-50G
FLUO005112-250GEA
0
GBP
FLUO005112-250G
FLUO005112-50G
Cyclopropyl methyl ketone
Cyclopropyl methyl ketone
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 114 Melting Pt: –68 °C Density: 0.899 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001297 CAS Number: 765-43-5 EINECS: 212-146-4 UN: 1992 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...