4-Bromophenyl methyl sulfone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003695-5G 003695-25G
FLUO003695-5GEA 0 GBP
FLUO003695-5G FLUO003695-25G
4-Bromophenyl methyl sulfone
4-Bromophenyl methyl sulfone
Formula: C₇H₇BrO₂S
MDL Number: MFCD00025065
CAS Number: 3466-32-8
EINECS: 222-421-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR