Bis(4-fluorophenyl)sulphone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorophenyl sulphone, 4,4'-Difluoro diphenyl sulphone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)

002477-25G 002477-100G
FLUO002477-25GEA 0 GBP
FLUO002477-25G FLUO002477-100G
Bis(4-fluorophenyl)sulphone
Bis(4-fluorophenyl) sulphone
Formula: C₁₂H₈F₂O₂S
MDL Number: MFCD00000350
CAS Number: 383-29-9
EINECS: 206-847-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR