4-Fluorophenylhydrazinium chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

001712-25G 001712-100G 001712-250G
FLUO001712-25GEA 0 GBP
FLUO001712-25G FLUO001712-100G FLUO001712-250G
4-Fluorophenylhydrazinium chloride
4-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MDL Number: MFCD00012942
CAS Number: 823-85-8
EINECS: 212-521-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR