5-Bromo-2-fluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

009413-25G 009413-100G
FLUO009413-25GEA 0 GBP
FLUO009413-25G FLUO009413-100G
5-Bromo-2-fluorobenzaldehyde
5-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 49 °C
Density: 1,71 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00070755
CAS Number: 93777-26-5
EINECS: 298-056-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR