3-Bromo-4-fluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
004208-25G
004208-100G
FLUO004208-25GEA
0
GBP
FLUO004208-25G
FLUO004208-100G
3-Bromo-4-fluorobenzaldehyde
3-Bromo-4-fluorobenzaldehyde
Formula:
C₇H₄BrFO Boiling Pt: 312 °C (1013 hPa) Melting Pt: 30…32 °C |
MDL Number:
MFCD00042186 CAS Number: 77771-02-9 EINECS: 278-764-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...