2-Bromo-4-chlorobenzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0994-5G OT-0994-25G
COBBOT-0994-5GEA 0 GBP
COBBOT-0994-5G COBBOT-0994-25G
2-Bromo-4-chlorobenzaldehyde
2-Bromo-4-chlorobenzaldehyde
Formula: C₇H₄BrClO
MDL Number: MFCD09056792
CAS Number: 84459-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR