2-Bromo-5-chlorobenzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0990-5G OT-0990-25G
COBBOT-0990-5GEA 0 GBP
COBBOT-0990-5G COBBOT-0990-25G
2-Bromo-5-chlorobenzaldehyde
2-Bromo-5-chlorobenzaldehyde
Formula: C₇H₄BrClO
MDL Number: MFCD00462870
CAS Number: 174265-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR