Methyl 2-amino-4,5-dimethoxybenzoate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 4,5-dimethoxyanthranilate, 2-Amino-4,5-dimethoxy benzoate, Methyl 2-amino-4,5-dimethoxybenzoate, 2-Amino-4,5-dimethoxybenzoic acid methyl ester
CA-4968-5G
CA-4968-1G
COBBCA-4968-5GEA
0
GBP
COBBCA-4968-5G
COBBCA-4968-1G
Methyl 2-amino-4,5-dimethoxybenzoate
Methyl 2-amino-4,5-dimethoxybenzoate
Formula:
C₁₀H₁₃NO₄ MW: 211.22 g/mol |
MDL Number:
MFCD00014904 CAS Number: 26759-46-6 EINECS: 247-967-7 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...