Methyl 2-bromo-4-methoxybenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-methoxybenzoic acid methyl ester, Methyl 2-bromo-4-methoxybenzoate

CA-4679-5G CA-4679-1G
COBBCA-4679-5GEA 0 GBP
COBBCA-4679-5G COBBCA-4679-1G
Methyl 2-bromo-4-methoxybenzoate
Methyl 2-bromo-4-methoxybenzoate
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD12026001
CAS Number: 17100-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR