Methyl 3-fluoro-4-methoxybenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-fluoro-4-methoxybenzoate, 3-Fluoro-4-methoxybenzoic acid methyl ester

CA-4659-1G CA-4659-5G
COBBCA-4659-1GEA 0 GBP
COBBCA-4659-1G COBBCA-4659-5G
Methyl 3-fluoro-4-methoxybenzoate
Methyl 3-fluoro-4-methoxybenzoate
Formula: C₉H₉FO₃
MW: 184.17 g/mol
MDL Number: MFCD00215839
CAS Number: 369-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR