Methyl 4-bromo-3-methoxybenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-3-methoxybenzoic acid methyl ester, Methyl 4-bromo-3-methoxybenzoate

CA-4192-5G CA-4192-1G
COBBCA-4192-5GEA 0 GBP
COBBCA-4192-5G COBBCA-4192-1G
Methyl 4-bromo-3-methoxybenzoate
Methyl 4-bromo-3-methoxybenzoate
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD08436018
CAS Number: 17100-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR