L-Cycloserine
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-(-)-Cycloserine
, L-4-Aminoisoxazolidin-3-one
, (S)-4-Aminoisoxazolidin-3-one
BACEF-1475.1000EA
1120
GBP
BACEF-1475.1000
BACEF-1475.0250
BACE4006516.0050
L-Cycloserine
L-Cycloserine
L-Cycloserine regulates sphingolipide metabolism by inhibiting the key enzyme serine palmitoyltransferase. Both cycloserine enantiomers inhibit 3-ketodihydrosphingosine synthetase (EC 2.3.1.50), though the L-form is the more potent inhibitor of the enzyme. Pérez-Sala et al. studied its inhibitory activity on pyruvate metabolism.
Formula:
C₃H₆N₂O₂ MW: 102.09 g/mol |
MDL Number:
MFCD00064324 CAS Number: 339-72-0 EINECS: 206-427-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...