Bromopentafluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-2,3,4,5,6-pentafluorobenzene

002832-50G 002832-250G 002832-1KG
FLUO002832-50GEA 0 GBP
FLUO002832-50G FLUO002832-250G FLUO002832-1KG
Bromopentafluorobenzene
Bromopentafluorobenzene
Formula: C₆BrF₅
MW: 246.96 g/mol
Boiling Pt: 137
Melting Pt: –31 °C
Density: 1.9503 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000287
CAS Number: 344-04-7
EINECS: 206-449-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR