2-(Biphenyl-4-yl)-2-propanol 95%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L01364.09
L01364.18
L01364.09EA
35.3
GBP
L01364.09
L01364.18
2-(Biphenyl-4-yl)-2-propanol 95%
Purity: 97%
CAS No.: 34352-74-4
Molecular Formula: C15H16O
Formula Weight: 212.29
Melting Point: 90-93°
Beilstein Reference: 1947575
Formula:
C₁₅H₁₆O Melting Pt: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014390 CAS Number: 34352-74-4 EINECS: 251-955-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...