2-Pentyl-1,3-propanediol ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Pentylpropane-1,3-diol, 2-Amyl-1,3-propanediol

B21800.03 B21800.06
B21800.03EA 19.8 GBP
B21800.03 B21800.06
2-Pentyl-1,3-propanediol ≥97%
2-Pentyl-1,3-propanediol

Purity: 97% CAS No.: 25462-23-1 Molecular Formula: CH3(CH2)4CH(CH2OH)2 Formula Weight: 146.23 MDL No.: MFCD00801123

Formula: C₈H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00801123
CAS Number: 25462-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR