Magnesium 2,4-pentanedionate hydrate ≥98% (metals basis)
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
12532.18
12532.30
12532.09
12532.18EA
57.1
GBP
12532.18
12532.30
12532.09
Magnesium 2,4-pentanedionate hydrate ≥98% (metals basis)
Magnesium 2,4-pentanedionate hydrate
Purity: 98+%
CAS No.: 14024-56-7
Molecular Formula: C10H14MgO4.xH2O
Formula Weight: 222.52(anhy)
Physical Form: Powder
Appearance: White
Melting Point: 260° dec.
MDL No.: MFCD00150168
Formula:
C₁₀H₁₄MgO₄·xH₂O Melting Pt: 260 °C (dec.) Storage Temperature: Ambient |
CAS Number:
67032-77-3 EINECS: 237-857-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...