1-Bromo-4-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromofluorobenzene

01225.500G 001225-1KG 001225-500G
FLUO01225.500GEA 0 GBP
FLUO01225.500G FLUO001225-1KG FLUO001225-500G
1-Bromo-4-fluorobenzene
1-Bromo-4-fluorobenzene
Formula: C₆H₄BrF
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1.4946 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00000342
CAS Number: 460-00-4
EINECS: 207-300-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR