1-Bromo-3-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromofluorobenzene

01224.100G 01224.500G 001224-100G 001224-500G
FLUO01224.100GEA 0 GBP
FLUO01224.100G FLUO01224.500G FLUO001224-100G FLUO001224-500G
1-Bromo-3-fluorobenzene
1-Bromo-3-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.7081 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000326
CAS Number: 1073-06-9
EINECS: 214-023-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR