1-Bromo-3,5-difluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3,5-Difluorobromobenzene
001416-500G
001416-250G
001416-100G
FLUO001416-500GEA
0
GBP
FLUO001416-500G
FLUO001416-250G
FLUO001416-100G
1-Bromo-3,5-difluorobenzene
1-Bromo-3,5-difluorobenzene
Formula:
C₆H₃BrF₂ MW: 192.99 g/mol Boiling Pt: 140 °C (1013 hPa) Melting Pt: –27 °C Density: 1.676 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010305 CAS Number: 461-96-1 EINECS: 416-710-2 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...