4-Bromo-1,2-difluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001118-100G
001118-250G
001118-500G
FLUO001118-100GEA
0
GBP
FLUO001118-100G
FLUO001118-250G
FLUO001118-500G
4-Bromo-1,2-difluorobenzene
4-Bromo-1,2-difluorobenzene
Formula:
C₆H₃BrF₂ Boiling Pt: 150 °C (1013 hPa) Melting Pt: –4 °C Density: 1.7 g/cm³ (20 °C) Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000304 CAS Number: 348-61-8 EINECS: 206-481-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...